MilliporeSigma
  • Mineralocorticoid receptor negatively regulates angiogenesis through repression of STAT3 activity in endothelial cells.
Mineralocorticoid receptor negatively regulates angiogenesis through repression of STAT3 activity in endothelial cells.
The Journal of pathology (2019-03-23)
Xiao-Jun Zheng, Yuan Liu, Wu-Chang Zhang, Yan Liu, Chao Li, Xue-Nan Sun, Yu-Yao Zhang, Jie Xu, Xinquan Jiang, Lanjing Zhang, Wei Yang, Sheng-Zhong Duan
ABSTRACT
The mineralocorticoid receptor (MR) plays important roles in cardiovascular pathogenesis. The function of MR in angiogenesis is still controversial. This study aimed to explore the role of endothelial MR in angiogenesis and to delineate the underlying mechanism. Endothelial-hematopoietic MR knockout (EMRKO) mice were generated and subjected to hindlimb ischemia and injection of melanoma cells. Laser Doppler measurements showed that EMRKO mice had improved blood flow recovery and increased vessel density in ischemic limbs. In addition, EMRKO accelerated growth and increased the vessel density of tumors. Matrigel implantation, aortic ring assays, and tube formation assays demonstrated that MRKO endothelial cells (ECs) manifested increased angiogenic potential. MRKO ECs also displayed increased migration ability and proliferation. MRKO and MR knockdown both upregulated gene expression, protein level, and phosphorylation of signal transducer and activator of transcription 3 (STAT3). Stattic, a selective STAT3 inhibitor, attenuated the effects of MRKO on tube formation, migration, and proliferation of ECs. At the molecular level, MR interacted with CCAAT enhancer-binding protein beta (C/EBPβ) to suppress the transcription of STAT3. Furthermore, interactions between MR and STAT3 blocked the phosphorylation of STAT3. Finally, stattic abolished the pro-angiogenic phenotype of EMRKO mice. Taken together, endothelial MR is a negative regulator of angiogenesis, likely in a ligand-independent manner. Mechanistically, MR downregulates STAT3 that mediates the impacts of MR deficiency on the angiogenic activity of ECs and angiogenesis. Targeting endothelial MR may be a potential pro-angiogenic strategy for ischemic diseases. © 2019 Pathological Society of Great Britain and Ireland. Published by John Wiley & Sons, Ltd.