MilliporeSigma
  • Induction of cystathionine gamma-lyase expression and metallothionein-1 S-sulfhydration alleviate cadmium-induced cell death in myoblast cells.
Induction of cystathionine gamma-lyase expression and metallothionein-1 S-sulfhydration alleviate cadmium-induced cell death in myoblast cells.
Ecotoxicology and environmental safety (2019-05-03)
Yanjie Zhang, Amr Ali, Zhuping Jin, Yanxi Pei, Guangdong Yang
ABSTRACT
Hydrogen sulfide (H2S), a multifunctional gasotransmitter, participates in a wide range of cellular signal transduction and pathophysiological processes. Cystathionine gamma-lyase (CSE) acts as a major H2S-generating enzyme in peripheral organs and tissues. As a cysteine-rich and heavy metal-binding protein, metallothionein-1 (MT-1) is known to protect cells from various environmental stresses. Here we demonstrated that exposure of cadmium (Cd) induced oxidative stress, depleted intracellular thiols, and stimulated apoptotic cell death in mouse myoblast cells. CSE expression and H2S production were significantly enhanced by Cd treatment. NaHS, a well-known H2S donor, at physiologically relevant concentration significantly alleviated Cd-induced damage in both myoblasts and mouse skeletal muscles. In contrast, down-regulation of CSE/H2S system deteriorated Cd-stimulated oxidative stress and cell death. Exposure of the cells to Cd lead to increased expressions of metal regulatory transcription factor 1 and MT-1, while siRNA-mediated MT-1 knockdown alleviated Cd-induced CSE expression and caused more oxidative stress and cell death. In addition, H2S post-translationally modified MT-1 by S-sulfhydration and stabilized zinc-protein complex. Taken together, these data suggest that CSE/H2S system would protect myoblasts and skeletal muscles from Cd-induced damage by S-sufhydrating MT-1.