MilliporeSigma
  • Prediction of DNA Integrity from Morphological Parameters Using a Single-Sperm DNA Fragmentation Index Assay.
Prediction of DNA Integrity from Morphological Parameters Using a Single-Sperm DNA Fragmentation Index Assay.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2019-08-14)
Yihe Wang, Jason Riordon, Tian Kong, Yi Xu, Brian Nguyen, Junjie Zhong, Jae Bem You, Alexander Lagunov, Thomas G Hannam, Keith Jarvi, David Sinton
ABSTRACT
Intracytoplasmic sperm injection is a popular form of in vitro fertilization, where single sperm are selected by a clinician and injected into an egg. Whereas clinicians employ general morphology-based guidelines to select the healthiest-looking sperm, it remains unclear to what extent an individual sperm's physical parameters correlate with the quality of internal DNA cargo-a measurement that cannot be obtained without first damaging the sperm. Herein, a single-cell DNA fragmentation index (DFI) assay is demonstrated, which combines the single-cell nature of the acridine orange test with the quantitative aspect of the sperm chromatin structure assay, to create a database of DFI-scored brightfield images. Two regression predictive models, linear and nonlinear regression, are used to quantify the correlations between individual sperm morphological parameters and DFI score (with model test r at 0.558 and 0.620 for linear and nonlinear regression models, respectively). The sample is also split into two categories of either relatively good or bad DFIs and a classification predictive model based on logistic regression is used to categorize sperm, resulting in a test accuracy of 0.827. Here, the first systematic study is presented on the correlation and prediction of sperm DNA integrity from morphological parameters at the single-cell level.