MilliporeSigma
  • 6,6-Spiroimine analogs of (-)-gymnodimine A: synthesis and biological evaluation on nicotinic acetylcholine receptors.
6,6-Spiroimine analogs of (-)-gymnodimine A: synthesis and biological evaluation on nicotinic acetylcholine receptors.
Organic & biomolecular chemistry (2011-10-26)
Leslie Duroure, Thierry Jousseaume, Rómulo Aráoz, Elvina Barré, Pascal Retailleau, Laurent Chabaud, Jordi Molgó, Catherine Guillou
ABSTRACT
Simple models of the spiroimine core of (-)-gymnodimine A have been synthesized in racemic and optically active forms. The quaternary carbon of the racemic spiroimines was created by Michael addition of a β-ketoester to acrolein, whereas the asymmetric allylic alkylation of the same β-ketoester was used to access the spiroimines in an enantioselective fashion. Both racemic and enantio-enriched mixtures were tested for their biological activities on Xenopus oocytes either expressing (human α4β2) or having incorporated (Torpedoα1(2)βγδ) nicotinic acetylcholine receptors (nAChRs). These spiroimine analogs of (-)-gymnodimine A inhibited acetylcholine-evoked nicotinic currents, but were less active than the phycotoxin. Our results reveal that the 6,6-spiroimine moiety is important for the blockade of nAChRs and support the hypothesis that it is one of the pharmacophores of this group of toxins.
Sigma-Aldrich
Vinylboronic acid pinacol ester, contains phenothiazine as stabilizer, 95%