MilliporeSigma
  • Liver-target nanotechnology facilitates berberine to ameliorate cardio-metabolic diseases.
Liver-target nanotechnology facilitates berberine to ameliorate cardio-metabolic diseases.
Nature communications (2019-05-02)
Hui-Hui Guo, Chen-Lin Feng, Wen-Xuan Zhang, Zhi-Gang Luo, Hong-Juan Zhang, Ting-Ting Zhang, Chen Ma, Yun Zhan, Rui Li, Song Wu, Zeper Abliz, Cong Li, Xiao-Lin Li, Xiao-Lei Ma, Lu-Lu Wang, Wen-Sheng Zheng, Yan-Xing Han, Jian-Dong Jiang
ABSTRACT
Cardiovascular and metabolic disease (CMD) remains a main cause of premature death worldwide. Berberine (BBR), a lipid-lowering botanic compound with diversified potency against metabolic disorders, is a promising candidate for ameliorating CMD. The liver is the target of BBR so that liver-site accumulation could be important for fulfilling its therapeutic effect. In this study a rational designed micelle (CTA-Mic) consisting of α-tocopheryl hydrophobic core and on-site detachable polyethylene glycol-thiol shell is developed for effective liver deposition of BBR. The bio-distribution analysis proves that the accumulation of BBR in liver is increased by 248.8% assisted by micelles. Up-regulation of a range of energy-related genes is detectable in the HepG2 cells and in vivo. In the high fat diet-fed mice, BBR-CTA-Mic intervention remarkably improves metabolic profiles and reduces the formation of aortic arch plaque. Our results provide proof-of-concept for a liver-targeting strategy to ameliorate CMD using natural medicines facilitated by Nano-technology.