MilliporeSigma
  • Anti-HSP70 autoantibody formation by B-1 cells in Toxoplasma gondii-infected mice.
Anti-HSP70 autoantibody formation by B-1 cells in Toxoplasma gondii-infected mice.
Infection and immunity (2000-08-19)
M Chen, F Aosai, H S Mun, K Norose, H Hata, A Yano
ABSTRACT
Formation of anti-Toxoplasma gondii HSP70 (TgHSP70) antibody cross-reactive to mouse HSP70 (mHSP70) was observed in the sera of BALB/c (a resistant strain) and C57BL/6 (B6; a susceptible strain) mice after peroral infection with T. gondii cysts of the Fukaya strain. The levels of anti-mHSP70 immunoglobulin G (IgG) autoantibody production in B6 mice were higher than those in BALB/c mice. The isotype and subclass of IgG of anti-TgHSP70 monoclonal antibodies cross-reactive to mHSP70 were mu and gamma3. Anti-mHSP70 autoantibody in T. gondii-infected BALB/c and B6 mice was shown to be produced by the CD5(+) subset of B cells (B-1a cells) but not by conventional B cells (B-2 cells). The epitopes recognized by anti-mHSP70 autoantibody were located primarily in the C-terminal fragment of mHSP70.