MilliporeSigma
  • Highly dispersed MoO(x) on carbon nanotube as support for high performance Pt catalyst towards methanol oxidation.
Highly dispersed MoO(x) on carbon nanotube as support for high performance Pt catalyst towards methanol oxidation.
Chemical communications (Cambridge, England) (2011-06-28)
Zhi Ming Cui, San Ping Jiang, Chang Ming Li
ABSTRACT
HPMo self-assembly on carbon nanotubes followed by decomposition is used to fabricate highly dispersed MoO(x)/CNTs as a support for high performance of a Pt catalyst towards methanol oxidation.
Product Description
Supelco
Phosphomolybdic acid hydrate, ≥99.99% trace metals basis