MilliporeSigma
  • FL-926-16, a novel bioavailable carnosinase-resistant carnosine derivative, prevents onset and stops progression of diabetic nephropathy in db/db mice.
FL-926-16, a novel bioavailable carnosinase-resistant carnosine derivative, prevents onset and stops progression of diabetic nephropathy in db/db mice.
British journal of pharmacology (2017-10-21)
Carla Iacobini, Stefano Menini, Claudia Blasetti Fantauzzi, Carlo M Pesce, Andrea Giaccari, Enrica Salomone, Annunziata Lapolla, Marica Orioli, Giancarlo Aldini, Giuseppe Pugliese
ABSTRACT
The advanced glycation end products (AGEs) participate in the pathogenesis of diabetic nephropathy (DN) by promoting renal inflammation and injury. L-carnosine acts as a quencher of the AGE precursors reactive carbonyl species (RCS), but is rapidly inactivated by carnosinase. In this study, we evaluated the effect of FL-926-16, a carnosinase-resistant and bioavailable carnosine derivative, on the onset and progression of DN in db/db mice. Adult male db/db mice and coeval db/m controls were left untreated or treated with FL-926-16 (30 mg·kg-1 body weight) from weeks 6 to 20 (prevention protocol) or from weeks 20 to 34 (regression protocol). In the prevention protocol, FL-926-16 significantly attenuated increases in creatinine (-80%), albuminuria (-77%), proteinuria (-75%), mean glomerular area (-34%), fractional (-40%) and mean (-42%) mesangial area in db/db mice. This protective effect was associated with a reduction in glomerular matrix protein expression and cell apoptosis, circulating and tissue oxidative and carbonyl stress, and renal inflammatory markers, including the NLRP3 inflammasome. In the regression protocol, the progression of DN was completely blocked, although not reversed, by FL-926-16. In cultured mesangial cells, FL-926-16 prevented NLRP3 expression induced by RCS but not by the AGE Nε -carboxymethyllysine. FL-926-16 is effective at preventing the onset of DN and halting its progression in db/db mice by quenching RCS, thereby reducing the accumulation of their protein adducts and the consequent inflammatory response. In a future perspective, this novel compound may represent a promising AGE-reducing approach for DN therapy.
Sigma-Aldrich
Anti-NLRP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution