MilliporeSigma
  • Open-tubular capillary electrochromatographic determination of ten sulfonamides in tap water and milk by a metal-organic framework-coated capillary column.
Open-tubular capillary electrochromatographic determination of ten sulfonamides in tap water and milk by a metal-organic framework-coated capillary column.
Electrophoresis (2018-05-26)
Xuan Wang, Nengsheng Ye, Xiaoyu Hu, Qingye Liu, Jian Li, Lin Peng, Xiaotong Ma
ABSTRACT
In this study, a metal-organic framework (MOF), [Mn(cam)(bpy)], was synthesized and characterized by thermogravimetric analysis, scanning electron microscopy, and Fourier transform infrared spectrometry. An open-tubular capillary column was fabricated from [Mn(cam)(bpy)] via the amide coupling method. Ten types of sulfonamides were separated through the fabricated capillary column, which showed a good limits of detection (<0.07 μg/mL) and linear ranges (1-100 or 5-100 μg/mL) with a high correlation coefficients (R2 > 0.9987). The intra-day, inter-day and column-to-column relative standard deviations (RSDs) in the migration times ranged from 0.44 to 4.87%, and the peak area RSDs ranged from 0.80 to 7.28%. The developed capillary electrochromatography method can be successfully utilized for the determination of sulfonamides in tap water and milk samples.
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%