MilliporeSigma
  • Hepcidin, an Iron Regulatory Hormone of Innate Immunity, is Differentially Expressed in Premature Fetuses with Early-Onset Neonatal Sepsis.
Hepcidin, an Iron Regulatory Hormone of Innate Immunity, is Differentially Expressed in Premature Fetuses with Early-Onset Neonatal Sepsis.
American journal of perinatology (2018-03-14)
Sammy M Tabbah, Catalin S Buhimschi, Katherine Rodewald-Millen, Christopher R Pierson, Vineet Bhandari, Philip Samuels, Irina A Buhimschi
ABSTRACT
Hepcidin, a mediator of innate immunity, binds the iron exporter ferroportin, leading to functional hypoferremia through intracellular iron sequestration. We explored hepcidin-ferroportin interactions in neonates clinically diagnosed with early-onset neonatal sepsis (EONS). Hepcidin and interleukin (IL)-6 were quantified by enzyme-linked immunosorbent assay (ELISA) in 92 paired cord blood-maternal blood samples in the following groups: "Yes" EONS (n = 41, gestational age [GA] 29 ± 1 weeks) and "No" EONS (n = 51, GA 26 ± 1 weeks). Placental hepcidin and ferroportin expression were evaluated by immunohistochemistry and real-time-polymerase chain reaction (RT-PCR). Liver hepcidin and ferroportin expression patterns were ascertained in autopsy specimens of neonates (n = 8) who died secondary to culture-proven sepsis. Cord blood hepcidin was significantly elevated (GA corrected, p = 0.018) and was positively correlated with IL-6 (r = 0.379, p = 0.001) in EONS. Hepcidin localized at syncytiotrophoblast and fetal vascular endothelium. Placental ferroportin, but not hepcidin mRNA correlated with cord blood hepcidin levels (r = 0.46, p = 0.039) and funisitis severity (r = 0.50, p = 0.018). Newborns who died from sepsis (n = 4) had higher hepatic hepcidin and iron sequestration, but lower ferroportin staining than those who died of nonsepsis causes (n = 4). Premature fetuses with EONS have elevated circulating hepcidin, likely related to lower placenta and liver ferroportin expression. Fetal hepcidin-ferroportin interaction appears to play a role in EONS pathophysiology independent of maternal response to intrauterine inflammation.
Sigma-Aldrich
IgG from goat serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder