MilliporeSigma
  • Time and temperature dependent adsorption-desorption behaviour of pretilachlor in soil.
Time and temperature dependent adsorption-desorption behaviour of pretilachlor in soil.
Ecotoxicology and environmental safety (2018-06-08)
Paawan Kaur, Pervinder Kaur
ABSTRACT
Understanding and quantifying the adsorption-desorption behaviour of herbicide in soil is imperative for predicting their fate and transport in the environment. In the present study, the effect of time and temperature on the adsorption-desorption behaviour of pretilachlor in soils was investigated using batch equilibration technique. The adsorption-desorption kinetics of pretilachlor in soils was two step process and was well described by pseudo-second-order kinetic model. Freundlich model accurately predicted the sorption behaviour of pretilachlor. The adsorption-desorption of pretilachlor varied significantly with the concentration, temperature and properties of soil viz. organic matter and clay content. All the studied soils had non-linear slopes (n < 1) and degree of nonlinearity increased with increase in clay, organic matter content and temperature (p < 0.05). Desorption of pretilachlor was hysteretic in studied soils and hysteresis coefficient varied from 0.023 to 0.275. Thermodynamic analysis showed that pretilachlor adsorption onto soils was a feasible, spontaneous and endothermic process which becomes more favourable at high temperature. It could be inferred that the adsorption of pretilachlor on soils was physical in nature.
Product Description
Supelco
Pretilachlor, PESTANAL®, analytical standard