MilliporeSigma
  • A new insight into the oxidative mechanism of caffeine and related methylxanthines in aprotic medium: May caffeine be really considered as an antioxidant?
A new insight into the oxidative mechanism of caffeine and related methylxanthines in aprotic medium: May caffeine be really considered as an antioxidant?
Biochimica et biophysica acta. General subjects (2018-05-16)
Rita Petrucci, Giuseppe Zollo, Antonella Curulli, Giancarlo Marrosu
ABSTRACT
Antioxidant properties have been recently suggested for caffeine that seems showing protective effects against damages caused by oxidative stress. In particular, a HO scavenging activity has been ascribed to caffeine. Even if the oxidation of caffeine has been widely studied, the antioxidant mechanism is still far to be understood. The electrochemical behavior of caffeine, theobromine and theophylline was studied in aprotic medium by cyclic voltammetry and electrolysis in UV-vis cell; a computational analysis of the molecular structures based on the Density Functional Theory was performed; the reactivity of all substrates towards lead dioxide, superoxide and galvinoxyl radical was followed by UV-vis spectrophotometry. Results supported the mono-electronic oxidation of the C4C5 bond for all substrates at high oxidation potentials, the electron-transfer process leading to a radical cation or a neutral radical according to the starting methylxanthine N7-substituted (caffeine and theobromine) or N7-unsubstituted (theophylline), respectively. A different following chemical fate might be predicted for the radical cation or the neutral radical. No interaction was evidenced towards the tested reactive oxygen species. No reactivity via H-atom transfer was evidenced for all studied compounds, suggesting that an antiradical activity should be excluded. Some reactivity only with strong oxidants could be predicted via electron-transfer. The acclaimed HO scavenging activity should be interpreted in these terms. The study suggested that CAF might be hardly considered an antioxidant. Beyond the experimental methods used, the discussion of the present results might provide food for thought to the wide audience working on antioxidants.
Sigma-Aldrich
Dicyclohexano-18-crown-6, 98%
Supelco
Theobromine, analytical standard