MilliporeSigma
  • A novel mediator of class II gene transcription with homology to viral immediate-early transcriptional regulators.
A novel mediator of class II gene transcription with homology to viral immediate-early transcriptional regulators.
Cell (1994-08-12)
M Kretzschmar, K Kaiser, F Lottspeich, M Meisterernst
ABSTRACT
Our investigations of mammalian class II gene transcription resulted in identification, purification, and cloning of the corresponding cDNA of a cellular factor (p15) that mediates the effects of several distinct activators on transcription in vitro. Functional deletion analyses revealed a bipartite structure of p15 comprising an amino-terminal regulatory domain and a carboxy-terminal cryptic DNA-binding domain. We provide evidence that activity of p15 is controlled by protein kinases that target the regulatory domain. Structural and functional similarities, including sequence homology to domains essential for cofactor function, cofactor activity, promiscuity with respect to transcriptional activators, and interactions with components of the basal transcription machinery, relate this novel cellular cofactor to viral immediate-early transcriptional regulators.
Sigma-Aldrich
PC4, F77P mutant human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
PC4, serine mutations human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)