MilliporeSigma
Interleukin-2: inception, impact, and implications.
Science (New York, N.Y.) (1988-05-27)
K A Smith
ABSTRACT
Interleukin-2 (IL-2), the first of a series of lymphocytotrophic hormones to be recognized and completely characterized, is pivotal for the generation and regulation of the immune response. A T lymphocyte product, IL-2 also stimulates T cells to undergo cell cycle progression via a finite number of interactions with its specific membrane receptors. Because T cell clonal proliferation after antigen challenge is obligatory for immune responsiveness and immune memory, the IL-2-T cell system has opened the way to a molecular understanding of phenomena that are fundamental to biology, immunology, and medicine.
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture