Skip to Content
MilliporeSigma
(Z)-Guggulsterone ≥89% (HPLC), powder

(Z)-Guggulsterone

(Z)-Guggulsterone ≥89% (HPLC), powder
Synonym(s):
(17Z)-Pregna-4,17(20)-diene-3,16-dione, 4,17(20)-trans-Pregnadiene-3,16-dione
Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing