Skip to Content
MilliporeSigma
TSQ ≥98% (HPLC)

TSQ

TSQ ≥98% (HPLC)
Synonym(s):
N-(6-Methoxy-8-quinolinyl)-4-methylbenzenesulfonamide
Empirical Formula (Hill Notation):
C17H16N2O3S
CAS Number:
Molecular Weight:
328.39

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing