
Soyasaponin Bb

Synonym(s):
Soyasaponin I, SCM 3B, Soyasaponin Bb
Empirical Formula (Hill Notation):
C48H78O18
CAS Number:
Molecular Weight:
943.12
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1