Oxyquinoline sulfate
Synonym(s):
8-Quinolinol hemisulfate salt, 8-Hydroxyquinoline hemisulfate salt
Empirical Formula (Hill Notation):
C18H14N2O2 · H2SO4
CAS Number:
Molecular Weight:
388.39
Products
Shipping
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing