Skip to Content
MilliporeSigma
Monoclonal Anti-Actin, α-Smooth Muscle clone 1A4, ascites fluid

Monoclonal Anti-Actin, α-Smooth Muscle

Monoclonal Anti-Actin, α-Smooth Muscle clone 1A4, ascites fluid
Synonym(s):
SMA

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Application
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing