Skip to Content
MilliporeSigma
5-(Hydroxymethyl)furfural analytical standard

5-(Hydroxymethyl)furfural

5-(Hydroxymethyl)furfural analytical standard
Synonym(s):
5-Hydroxymethyl-2-furaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethyl-2-furancarboxaldehyde, HMF
Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing