Skip to Content

T154

Tityustoxin-Kα from Tityus serrulatus (Brazilian scorpion) venom

lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

solubility

H2O: soluble

storage temp.

−20°C

Gene Information

Biochem/physiol Actions

Potent and selective voltage-gated non-inactivating (delayed-rectifier) K+ channel blocker.

Legal Information

Sold under license from the University of Maryland.

Disclaimer

Hygroscopic, photosensitive. Only plastic tubes should be used with this product.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library