Skip to Content
MilliporeSigma

S2877

Tin(II) fluoride

Synonym(s):

Difluorostannylene, Stannous fluoride, Tin bifluoride, Tin difluoride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
F2Sn
CAS Number:
Molecular Weight:
156.71
EC Number:
231-999-3
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


density

4.57 g/mL at 25 °C (lit.)

SMILES string

F[SnH2]F

InChI

1S/2FH.Sn/h2*1H;/q;;+2/p-2

InChI key

ANOBYBYXJXCGBS-UHFFFAOYSA-L



signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library