Skip to Content
MilliporeSigma

R8907

RPI-1

≥98% (HPLC)

Synonym(s):

1,3-dihydro-5,6-dimethoxy-3-[(4-hydroxyphenyl)methylene]-H-indol-2-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H15NO4
CAS Number:
Molecular Weight:
297.31
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

orange

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

COc1cc2NC(=O)C(=Cc3ccc(O)cc3)c2cc1OC

InChI

1S/C17H15NO4/c1-21-15-8-12-13(7-10-3-5-11(19)6-4-10)17(20)18-14(12)9-16(15)22-2/h3-9,19H,1-2H3,(H,18,20)

InChI key

JGSMCYNBVCGIHC-UHFFFAOYSA-N

Biochem/physiol Actions

RPI-1 is a competitive, potent ATP-dependent Ret kinase inhibitor.
RPI-1 is a competitive, potent ATP-dependent Ret kinase inhibitor. Recently it was discover that the compound also inhibits c-Met. Increased tumorigenicity, motility, and invasiveness have been described as biological consequences of HGF/Met deregulation in tumor cells, thus not surprisingly RPI-1 treatment of H460 cells resulted in a strong reduction of both colony number and size (IC50 = 24.5 + 0.5 microM). Compound is also active at mouse NSCLC H460 xenograft tumor and metastasis model. Mechanistically RPI-1 inhibits Met phosphorylation at Tyr1234/Tyr1235, known to activate the intrinsic kinase activity. It appears that " Ret/ptc1 cross talks with Met at transcriptional and signaling levels and promotes ?-catenin transcriptional activity to drive thyrocyte neoplastic transformation". It appears that we do not have anything specific in Ret area. Handbook lists RPI-1 as Ret inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library