Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C18H21N3O5S
CAS Number:
Molecular Weight:
391.44
UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:
form
solid
storage temp.
−20°C
SMILES string
CC(C)(C)Oc1ccc(CC(NSc2ncccc2N(=O)=O)C(O)=O)cc1
InChI
1S/C18H21N3O5S/c1-18(2,3)26-13-8-6-12(7-9-13)11-14(17(22)23)20-27-16-15(21(24)25)5-4-10-19-16/h4-10,14,20H,11H2,1-3H3,(H,22,23)
InChI key
DTMLYWDPCJWGPQ-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.