Skip to Content
MilliporeSigma

F9132

Fmoc-Asp(OtBu)-OH

Synonym(s):

Fmoc-L-aspartic acid 4-tert-butyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H25NO6
CAS Number:
Molecular Weight:
411.45
EC Number:
276-251-7
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
3635671
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)C[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C23H25NO6/c1-23(2,3)30-20(25)12-19(21(26)27)24-22(28)29-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,12-13H2,1-3H3,(H,24,28)(H,26,27)/t19-/m0/s1

InChI key

FODJWPHPWBKDON-IBGZPJMESA-N



Still not finding the right product?

Explore all of our products under Fmoc-Asp(OtBu)-OH


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library