Skip to Content
MilliporeSigma

E5900

Anti-c-erbB-4 antibody, Mouse monoclonal

clone HER4-36, purified from hybridoma cell culture

Synonym(s):

Anti-HER-4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
HER4-36, monoclonal
Application:
ARR, IP
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HER4-36, monoclonal

form

buffered aqueous solution

mol wt

antigen 180-185 kDa

species reactivity

human

concentration

~2 mg/mL

technique(s)

immunoprecipitation (IP): 4-8 μg/test using RIPA lysate of cultured CB4 cells (CHO cell line transfected with erbB-4), microarray: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ERBB4(2066)

General description

Monoclonal Anti-c-erbB-4 (erb-b2 receptor tyrosine kinase 4) (HER-4) (mouse IgG1 isotype) is derived from the HER4-36 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c mouse immunized with a purified extracellular domain of human erbB-4 from transfected cells. The human ErbB-4 oncogene (also known as HER4) is a 180–185 kDa receptor that binds to a family of isoforms, collectively known as neuregulins. This gene belongs to epidermal growth factor receptor family and is expressed mainly in several breast carcinoma cell lines, heart, brain, cerebellum, pituitary and in normal skeletal muscle.

Immunogen

extracellular domain of human erbB-4 from transfected cells.

Application

Anti-c-erbB-4 antibody, Mouse monoclonal has been used in immunoprecipitation. It has also been used in studies on receptor turnover, phosphorylation, ligand binding and induction of differentiation of cultured breast cancer cells.
Monoclonal anti-c-erbB-4 antibody can be used in immunoprecipitation (4-8 μg/test) using RIPA lysate of cultured CB4 cells (CHO cell line transfected with erbB-4). It can also be used in microarray.

Biochem/physiol Actions

The epidermal growth factor receptor (ErbB/HER) family of transmembrane receptor tyrosine kinases significantly participates in defining cell lineage during mesenchyme-epithelial inductive processes and in the neuro-muscular and glia-Schwann cells interactions. It plays a crucial role as a cell surface receptor for neuregulins. It can also bind with erbB -2 to forms heterodimers and can modulate ligand affinity. The catalytic domain is important for the intrinsic tyrosine kinase activity and shows greater degree of homology. Certain ligands and receptors of the family contribute to a relatively virulent phenotype of some human tumors; most notable are carcinomas of secretory epithelia. ErbB-4 also binds to related group of molecules, termed neuregulin 2 (NRG2)(which also binds to erbB-3) and NRG3 (which binds exclusively to erbB-4). Two other ligands, betacellulin and the heparin-binding EGF-like growth factor, bind to both erbB-1 and erbB-4. ErbB-4 forms heterodimers with erbB-2, resulting in the modulation of its ligand affinity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library