Skip to Content

D9756

N-Dansyl-DL-serine cyclohexylammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H18N2O5S · C6H13N
CAS Number:
Molecular Weight:
437.55
PubChem Substance ID:
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C

SMILES string

NC1CCCCC1.CN(C)c2cccc3c(cccc23)S(=O)(=O)NC(CO)C(O)=O

InChI

1S/C15H18N2O5S.C6H13N/c1-17(2)13-7-3-6-11-10(13)5-4-8-14(11)23(21,22)16-12(9-18)15(19)20;7-6-4-2-1-3-5-6/h3-8,12,16,18H,9H2,1-2H3,(H,19,20);6H,1-5,7H2

InChI key

NEVMXASXEDTEQO-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library