Skip to Content

A8157

α-Adenosine

≥85%

Synonym(s):

9-α-Ribofuranosyladenine, 9-(α-D-Ribofuranosyl)adenine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
UNSPSC Code:
41106305
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥85%

form

powder

mp

205-207 °C (lit.)

solubility

1 N NH4OH: 50 mg/mL, clear to slightly hazy, colorless to yellow

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@H]3O[C@H](CO)[C@@H](O)[C@H]3O

InChI

1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7-,10+/m1/s1

InChI key

OIRDTQYFTABQOQ-CRKDRTNXSA-N



Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library