Skip to Content
MilliporeSigma

60354

Potassium phosphate dibasic

BioUltra, anhydrous, ≥99.0% (T)

Synonym(s):

Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
PubChem Substance ID:
eCl@ss:
38070212
UNSPSC Code:
12161700
EC Number:
231-834-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioUltra

assay

≥99.0% (T)

impurities

insoluble matter, passes filter test

pH

8.7-9.3 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤10 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤5000 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.20, λ: 280 nm Amax: ≤0.15

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

Other Notes

Component of a wide variety of media for the culture of microorganisms. Component of Dulbecco′s phosphate buffered saline for cell culture in which it maintains the pH and supplies essential phosphate
Suitable alkalimetric standard for standardizing NaOH, KOH, Ba(OH)2.


Still not finding the right product?

Explore all of our products under Potassium phosphate dibasic


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library