Skip to Content

12827

Sodium benzoxazole-2-sulfonate

purum p.a., derivatization grade (for amines and amino acids by HPLC), ≥95.0% (HPLC)

Synonym(s):

Benzoxazole-2-sulfonic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H4NNaO4S
CAS Number:
Molecular Weight:
221.17
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

derivatization grade (for amines and amino acids by HPLC), purum p.a.

assay

≥95.0% (HPLC)

impurities

≤2% water

storage temp.

2-8°C

SMILES string

[Na+].[O-]S(=O)(=O)c1nc2ccccc2o1

Other Notes

Derivatizing agent for the UV or fluorometric determination of prim. or sec. amines and amino acids




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library