Skip to Content

VT001076

Legionella pneumophila serogroup 1 WDCM 00107  Vitroids

1,000-10,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

greener alternative

Greener Alternative

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

sustainability

Greener Alternative Product

application(s)

microbiology

greener alternative category

storage temp.

−20°C

General description

We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of our four categories of greener alternatives. Click here to view its DfS scorecard.

Other Notes

Strain History

< CIP, 1993 < C. Tram, Inst. Pasteur, Paris, France < ATCC < R.E. Weaver, CDC: strain Philadelphia 1 < J.E. McDade

Legal Information

Vitroids is a trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library