Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1768

High-purity iron

NIST® SRM® 1768

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

application(s)

pharmaceutical (small molecule)

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is a high-purity iron disk. SRM 1768 is intended for use in optical emission and X-ray spectrometric methods of analysis. A unit of SRM 1768 consists of a disk approximately 31 mm (l.2 in) in diameter and 19 mm (0.75 in) thick. For more information, plese refer to the COA and SDS.

SRM 1768_cert

SRM 1768_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service