Skip to Content
MilliporeSigma

55898

Conductivity Standard

conductivity 500000 μS/cm at 25 °C

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

concentration

2.5-10% HCl (T)

conductivity

495000-505000 μS/cm at 25 °C

density

1.011-1.047 g/cm3 at 20 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

single component solution

General description

Conductivity standard can generally be used for the calibration of conductivity sensors for conductivity measurements. The conductivity standard solution for conductivity measurements, is either sodium chloride or potassium chloride solution of a definite concentration.


Still not finding the right product?

Explore all of our products under Conductivity Standard


pictograms

CorrosionExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library