Skip to Content
MilliporeSigma

429481

3,6-Dioctyloxy-1,2-benzenedicarbonitrile

97%

Synonym(s):

3,6-Dioctyloxyphthalonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[CH3(CH2)7O]2C6H2(CN)2
CAS Number:
Molecular Weight:
384.55
UNSPSC Code:
12171500
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

97%

mp

143-147 °C (lit.)

SMILES string

CCCCCCCCOc1ccc(OCCCCCCCC)c(C#N)c1C#N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library