Skip to Content
MilliporeSigma

19-0050

Magnesium

≥99.8%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Mg
CAS Number:
Molecular Weight:
24.31
PubChem Substance ID:
UNSPSC Code:
12352300
Beilstein/REAXYS Number:
4948473
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

1 mmHg ( 621 °C)

assay

≥99.8%

autoignition temp.

950 °F

reaction suitability

reagent type: reductant

availability

available only in Japan

resistivity

4.46 μΩ-cm, 20°C

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Magnesium


pictograms

Flame

signalword

Danger

hcodes

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

Hazard Classifications

Flam. Sol. 1 - Water-react. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library