Skip to Content
MilliporeSigma

Skip To

PGN003

PureGenome™ Next Generation Sequencing Library Amplifier

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352200
This product is sold exclusively by Millipore via offline channels. Contact Customer Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

Legal Information

PUREGENOME is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MBD0013MBD0014OLIGO
description

-

description

Suitable for PCR, sequencing and NGS, 10 ng/μL

description

Suitable for PCR, sequencing and NGS, ≥10 ng/μL

description

Design and Order Custom Oligos

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

-

Sigma-Aldrich PureGenome NGS Library Amplifier

-

-

-


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library