Skip to Content
MilliporeSigma

COMH93D5DA70

Combi-Blocks, Inc.

1-Dodecyl-3-methylimidazolium bis(trifluoromethanesulfonyl)imide

Synonym(s):

1-dodecyl-3-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C18H31O4N3S2F6
CAS Number:
Molecular Weight:
531.58
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C16H31N2.C2F6NO4S2/c1-3-4-5-6-7-8-9-10-11-12-13-18-15-14-17(2)16-18;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h14-16H,3-13H2,1-2H3;/q+1;-1

Inchi Key

RJCNRVFKCLDBEC-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library