Skip to Content

COM448658353

Combi-Blocks, Inc.

5-Bromo-2-chloromethylbenzofuran

Synonym(s):

5-bromo-2-(chloromethyl)-1-benzofuran

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H6O1Cl1Br1
CAS Number:
Molecular Weight:
245.5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H6BrClO/c10-7-1-2-9-6(3-7)4-8(5-11)12-9/h1-4H,5H2

Inchi Key

IGVKTJSZEQNSNH-UHFFFAOYSA-N

Purity

97%

Storage temp.

Refrigerated

Country of Origin

CN



Still not finding the right product?

Explore all of our products under 5-Bromo-2-chloromethylbenzofuran




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library