Skip to Content

COM448640348

Combi-Blocks, Inc.

3-Oxo-3-(6'-methoxylcarbonylpyridin-2-yl)propanenitrile

Synonym(s):

methyl 6-(cyanoacetyl)-2-pyridinecarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H8O3N2
CAS Number:
Molecular Weight:
204.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H8N2O3/c1-15-10(14)8-4-2-3-7(12-8)9(13)5-6-11/h2-4H,5H2,1H3

Inchi Key

MKVNIIBUDQRNHR-UHFFFAOYSA-N

Purity

95%

Storage temp.

Refrigerated

Country of Origin

CN



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library