Skip to Content
MilliporeSigma

COM448635670

Combi-Blocks, Inc.

3-Bromo-6-hydrazinyl-2-methylpyridine

Synonym(s):

3-bromo-6-hydrazino-2-methylpyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
202.05
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H8BrN3/c1-4-5(7)2-3-6(9-4)10-8/h2-3H,8H2,1H3,(H,9,10)

Inchi Key

WQIMBATZAFAPJX-UHFFFAOYSA-N

Storage temp.

Freezer

Purity

98%

Country of Origin

CN



Still not finding the right product?


MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?PY-7093



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library