Merck
CCR2
Mucosal immunology (2017-01-26)
J Liu, Y Xue, D Dong, C Xiao, C Lin, H Wang, F Song, T Fu, Z Wang, J Chen, H Pan, Y Li, D Cai, Z Li
ABSTRACT
Macrophages are distributed throughout the body and are crucial for the restoration of damaged tissues. However, their characteristics in the cornea and roles in the repair of corneal injures are unclear. Here we show that corneal macrophages can be classified as CCR2
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid