Merck
  • Modulation of chemical dermal absorption by 14 natural products: a quantitative structure permeation analysis of components often found in topical preparations.
Modulation of chemical dermal absorption by 14 natural products: a quantitative structure permeation analysis of components often found in topical preparations.
Cutaneous and ocular toxicology (2016-11-09)
Faqir Muhammad, Majid Jaberi-Douraki, Damião Pergentino de Sousa, Jim E Riviere
ABSTRACT
A large number of cosmetics and topical pharmaceuticals contain compounds of natural origin. There is a rising concern if these compounds can interact with the activity of other topically applied components in these formulations. The current study demonstrates modulation of dermal absorption of model components often found in topical preparations (
Sigma-Aldrich
Menthyl acetate, 97%