Merck
  • Comparative study on fatty acid metabolism of early stages of two crustacean species: Artemia sp. metanauplii and Grapsus adscensionis zoeae, as live prey for marine animals.
Comparative study on fatty acid metabolism of early stages of two crustacean species: Artemia sp. metanauplii and Grapsus adscensionis zoeae, as live prey for marine animals.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2016-11-15)
Diana B Reis, Nieves G Acosta, Eduardo Almansa, Juan C Navarro, Douglas R Tocher, José P Andrade, António V Sykes, Covadonga Rodríguez
ABSTRACT
The present study compared the lipid composition and in vivo capability of Artemia sp. metanauplii (the main live prey used in aquaculture) and Grapsus adscensionis zoeae (as a wild zooplankton model) to metabolise unsaturated fatty acids. The two species were incubated in vivo with 0.3μM of individual [1-