Merck
Oncogenic Ras influences the expression of multiple lncRNAs.
Cytotechnology (2014-12-17)
Yojiro Kotake, Madoka Naemura, Kyoko Kitagawa, Hiroyuki Niida, Toshiyuki Tsunoda, Senji Shirasawa, Masatoshi Kitagawa
ABSTRACT
Recent ultrahigh-density tiling array and large-scale transcriptome analysis have revealed that large numbers of long non-coding RNAs (lncRNAs) are transcribed in mammals. Several lncRNAs have been implicated in transcriptional regulation, organization of nuclear structure, and post-transcriptional processing. However, the regulation of expression of lncRNAs is less well understood. Here, we show that the exogenous and endogenous expression of an oncogenic form of small GTPase Ras (called oncogenic Ras) decrease the expression of lncRNA ANRIL (antisense non-coding RNA in the INK4 locus), which is involved in the regulation of cellular senescence. We also show that forced expression of oncogenic Ras increases the expression of lncRNA PANDA (p21 associated ncRNA DNA damage activated), which is involved in the regulation of apoptosis. Microarray analysis demonstrated that expression of multiple lncRNAs fluctuated by forced expression of oncogenic Ras. These findings indicate that oncogenic Ras regulates the expression of a large number of lncRNAs including functional lncRNAs, such as ANRIL and PANDA.
Sigma-Aldrich
Anti-c-H-Ras (Ab-1) Mouse mAb (F235-1.7.1), liquid, clone F235-1.7.1, Calbiochem®