- A new "turn-on" chemodosimeter for Hg2+: ICT fluorophore formation via Hg(2+)-induced carbaldehyde recovery from 1,3-dithiane.
A new "turn-on" chemodosimeter for Hg2+: ICT fluorophore formation via Hg(2+)-induced carbaldehyde recovery from 1,3-dithiane.
Chemical communications (Cambridge, England) (2012-04-20)
Yuncong Chen, Chengcheng Zhu, Zhenghao Yang, Jing Li, Yang Jiao, Weijiang He, Junjie Chen, Zijian Guo
PMID22514021
ABSTRACT
A novel sensitive and specific Hg(2+) chemodosimeter, derived from 1',3'-dithiane-substituted 2,1,3-benzoxadiazole, displays "turn-on" fluorescent and colorimetric responses via an Hg(2+)-triggered aldehyde recovery reaction. Its potential to monitor Hg(2+) in living organisms has been demonstrated using zebrafish larvae.
Product Description
Pricing and availability is not currently available. Contact Technical Service