Merck
  • Targeting the proangiogenic VEGF-VEGFR protein-protein interface with drug-like compounds by in silico and in vitro screening.
Targeting the proangiogenic VEGF-VEGFR protein-protein interface with drug-like compounds by in silico and in vitro screening.
Chemistry & biology (2011-12-27)
Benoit Gautier, Maria A Miteva, Victor Goncalves, Florent Huguenot, Pascale Coric, Serge Bouaziz, Bili Seijo, Jean-François Gaucher, Isabelle Broutin, Christiane Garbay, Aurelien Lesnard, Sylvain Rault, Nicolas Inguimbert, Bruno O Villoutreix, Michel Vidal
ABSTRACT
Protein-protein interactions play a central role in medicine, and their modulation with small organic compounds remains an enormous challenge. Because it has been noted that the macromolecular complexes modulated to date have a relatively pronounced binding cavity at the interface, we decided to perform screening experiments over the vascular endothelial growth factor receptor (VEGFR), a validated target for antiangiogenic treatments with a very flat interface. We focused the study on the VEGFR-1 D2 domain, and 20 active compounds were identified. These small compounds contained a (3-carboxy-2-ureido)thiophen unit and had IC(50) values in the low micromolar range. The most potent compound inhibited the VEGF-induced VEGFR-1 transduction pathways. Our findings suggest that our best hit may be a promising scaffold to probe this macromolecular complex and for the development of treatments of VEGFR-1-dependent diseases.
Sigma-Aldrich
Tetrahydrothiophene, 99%