Merck
  • Synthesis of neurotensin(8-13)-phosphopeptide heterodimers via click chemistry.
Synthesis of neurotensin(8-13)-phosphopeptide heterodimers via click chemistry.
Bioorganic & medicinal chemistry letters (2010-05-11)
Susan Richter, Theres Ramenda, Ralf Bergmann, T Kniess, Joerg Steinbach, Jens Pietzsch, Frank Wuest
ABSTRACT
Two neurotensin(8-13)-containing peptide heterodimers were prepared via copper(I)-mediated click chemistry. The resulting peptide dimers could be obtained in 28-31% yield after HPLC purification. Neurotensin(8-13)-containing peptide dimers were used in an in vitro binding assay to determine binding affinity towards the neurotensin receptor-1 (NTR1). The determined IC(50) values of 8.3 microM and 0.7 microM indicate only very low binding affinity of the neurotensin(8-13)-containing peptide heterodimers towards the NTR1.
Sigma-Aldrich
Neurotensin Fragment 8-13 acetate salt, ≥97% (HPLC)