- The roles of dopamine transport inhibition and dopamine release facilitation in wake enhancement and rebound hypersomnolence induced by dopaminergic agents.
The roles of dopamine transport inhibition and dopamine release facilitation in wake enhancement and rebound hypersomnolence induced by dopaminergic agents.
Rebound hypersomnolence (RHS: increased sleep following increased wake) is a limiting side-effect of many wake-promoting agents. In particular, RHS in the first few hours following wake appears to be associated with dopamine (DA)-releasing agents, e.g., amphetamine, but whether it can also be produced by DA transporter (DAT) inhibition alone is unknown. In these studies, DA-releasing and DAT-inhibiting agents and their interaction were systematically examined for their ability to increase wake and induce RHS. Chronically implanted rats were evaluated in a blinded, pseudo-randomized design. 237 rats were used in these studies with 1 week between repeat tests. Animals were habituated overnight and dosed the next day, 5 h after lights on, with test agents. Sleep/wake activityand RHS were evaluated using EEG/EMG recording up to 22 h post dosing. In vitro dopamine release was evaluated in rat synaptosomes. At doses that produced equal increases in wake, DA-releasing (amphetamine, methamphetamine, phentermine) and several DAT-inhibiting agents (cocaine, bupropion, and methylphenidate) produced RHS during the first few hours after the onset of sleep recovery. However, other DAT-inhibiting agents (mazindol, nomifensine, GBR-12909, and GBR-12935) did not produce RHS. Combination treatment with amphetamine and nomifensine produced waking activity greater than the sum of their individual activities alone while ameliorating the amphetamine-like RHS. In rat synaptosomes, nomifensine reduced the potency of amphetamine to induce DA release approximately 270-fold, potentially explaining its action in ameliorating amphetamine-induced RHS. All DA releasing agents tested, and some DAT-inhibiting agents, produced RHS at equal wake-promoting doses. Thus amphetamine-like DA release appears sufficient for inducing RHS, but additional properties (pharmacologic and/or pharmacokinetic) evidently underlie RHS of other DAT inhibitors. Enhancing wake while mitigating RHS can be achieved by combining DAT-inhibiting and DA-releasing agents.