Merck
Polymorphism in iodotris(tri-p-tolylphosphine)silver(I).
Acta crystallographica. Section B, Structural science (2009-03-21)
Gertruida J S Venter, Andreas Roodt, Reinout Meijboom
ABSTRACT
The reaction of silver(I) iodide with tri(p-tolyl)phosphine in MeCN solution in 1:3 molar ratio yields a polymorph of the complex of the formula [AgI{P(4-MeC6H4)3}3], with the Ag atom in a distorted tetrahedral environment. A polymorphic structure of this complex (a) is compared with previously published crystal structures (b), determined at different temperatures. The two polymorphs are compared using r.m.s. overlay calculations as well as half-normal probability plots.
Sigma-Aldrich
Silver iodide, 99.999% trace metals basis
Sigma-Aldrich
Silver iodide, 99%