Merck
  • Aminocyclopentanols as sugar mimics. Synthesis from unsaturated bicyclic lactones by Overman rearrangement.
Aminocyclopentanols as sugar mimics. Synthesis from unsaturated bicyclic lactones by Overman rearrangement.
Organic & biomolecular chemistry (2007-09-20)
Marie Bøjstrup, Mette Fanefjord, Inge Lundt
ABSTRACT
Bicyclic cyclopentane lactones, prepared from bromodeoxyaldonolactones, were transformed into aminocyclopentanols with an Overman rearrangement as the key step. Two of the compounds prepared, 7 and 19, were found to be good inhibitors of jack bean alpha-mannosidase and beta-D-N-acetylglucosaminidase, respectively.
Sigma-Aldrich
Cyclopentanol, 99%